Assay Details
| Gene Symbol: SOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035543.1 | NP_001030620.1 | 1 - 2 | 159 | 68 | |
| NM_111057.4 | NP_186840.1 | 5 - 6 | 530 | 68 | ||
| GenBank mRNA | AB071965.1 | - | 5 - 6 | 530 | 68 | |
| AF360247.1 | - | 5 - 6 | 411 | 68 | ||
| AK226455.1 | - | 5 - 6 | 412 | 68 | ||
| AY133863.1 | - | 5 - 6 | 373 | 68 | ||
More Information
Set Membership: |
|

SOX
Genomic Map