Assay Details
| Gene Symbol: SOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035543.1 | NP_001030620.1 | 2 - 3 | 276 | 89 | |
| NM_111057.4 | NP_186840.1 | 6 - 7 | 647 | 89 | ||
| GenBank mRNA | AB071965.1 | - | 6 - 7 | 647 | 89 | |
| AF360247.1 | - | 6 - 7 | 528 | 89 | ||
| AK226455.1 | - | 6 - 7 | 529 | 89 | ||
| AY133863.1 | - | 6 - 7 | 490 | 89 | ||
More Information
Set Membership: |
|

SOX
Genomic Map