Assay Details
| Gene Symbol: SOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035543.1 | NP_001030620.1 | 3 - 4 | 380 | 76 | |
| NM_001084620.1 | NP_001078089.1 | 3 - 4 | 297 | 76 | ||
| NM_111057.4 | NP_186840.1 | 7 - 8 | 751 | 76 | ||
| GenBank mRNA | AB071965.1 | - | 7 - 8 | 751 | 76 | |
| AF360247.1 | - | 7 - 8 | 632 | 76 | ||
| AK226455.1 | - | 7 - 8 | 633 | 76 | ||
| AY133863.1 | - | 7 - 8 | 594 | 76 | ||
More Information
Set Membership: |
|

SOX
Genomic Map