Assay Details
| Gene Symbol: SOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035543.1 | NP_001030620.1 | 4 - 5 | 485 | 106 | |
| NM_001084620.1 | NP_001078089.1 | 4 - 5 | 402 | 106 | ||
| NM_111057.4 | NP_186840.1 | 8 - 9 | 856 | 106 | ||
| GenBank mRNA | AB071965.1 | - | 8 - 9 | 856 | 106 | |
| AF360247.1 | - | 8 - 9 | 737 | 106 | ||
| AK226455.1 | - | 8 - 9 | 738 | 106 | ||
| AY133863.1 | - | 8 - 9 | 699 | 106 | ||
More Information
Set Membership: |
|

SOX
Genomic Map