Assay Details
| Gene Symbol: SOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035543.1 | NP_001030620.1 | 5 - 6 | 586 | 67 | |
| NM_001084620.1 | NP_001078089.1 | 5 - 6 | 503 | 67 | ||
| NM_111057.4 | NP_186840.1 | 9 - 10 | 957 | 67 | ||
| GenBank mRNA | AB071965.1 | - | 9 - 10 | 957 | 67 | |
| AF360247.1 | - | 9 - 10 | 838 | 67 | ||
| AK226455.1 | - | 9 - 10 | 839 | 67 | ||
| AY133863.1 | - | 9 - 10 | 800 | 67 | ||
More Information
Set Membership: |
|

SOX
Genomic Map