Assay Details
| Gene Symbol: RAD23C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202864.1 | NP_001189793.1 | 4 - 5 | 517 | 61 | |
| NM_111121.4 | NP_186903.1 | 4 - 5 | 611 | 61 | ||
| NM_202482.3 | NP_974211.1 | 4 - 5 | 539 | 61 | ||
| GenBank mRNA | AB109196.1 | - | 4 - 5 | 348 | 61 | |
| AY039562.1 | - | 4 - 5 | 478 | 61 | ||
| AY113034.1 | - | 4 - 5 | 348 | 61 | ||
More Information
Set Membership: |
|

RAD23C
Genomic Map