Assay Details
| Gene Symbol: MCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337453.1 | NP_001326201.1 | 1 - 2 | 436 | 70 | |
| NM_111168.2 | NP_186948.1 | 1 - 2 | 425 | 70 | ||
| GenBank mRNA | EU598461.1 | - | 1 - 2 | 147 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001337453.1 transcripts.
Set Membership: |
|

MCC1
Genomic Map