Assay Details
| Gene Symbol: MCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111168.2 | NP_186948.1 | 2 - 3 | 566 | 148 | |
| GenBank mRNA | EU598461.1 | - | 2 - 3 | 288 | 148 | |
More Information
Set Membership: |
|

MCC1
Genomic Map