Assay Details
| Gene Symbol: MCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337453.1 | NP_001326201.1 | 2 - 3 | 994 | 97 | |
| NM_111168.2 | NP_186948.1 | 3 - 4 | 673 | 97 | ||
| GenBank mRNA | EU598461.1 | - | 3 - 4 | 395 | 97 | |
More Information
Set Membership: |
|

MCC1
Genomic Map