Assay Details
| Gene Symbol: XPO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111181.3 | NP_566193.2 | 31 - 32 | 3372 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XPO1B| Gene Symbol: XPO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111181.3 | NP_566193.2 | 31 - 32 | 3372 | 109 | |
Set Membership: |
|