Assay Details
| Gene Symbol: RIF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111242.3 | NP_187021.2 | 16 - 17 | 2133 | 78 | |
| GenBank mRNA | AF450480.1 | - | 16 - 17 | 1953 | 78 | |
More Information
Set Membership: |
|

RIF10
Genomic Map