Assay Details
| Gene Symbol: RIF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111242.3 | NP_187021.2 | 5 - 6 | 776 | 75 | |
| GenBank mRNA | AF450480.1 | - | 5 - 6 | 596 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIF10| Gene Symbol: RIF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111242.3 | NP_187021.2 | 5 - 6 | 776 | 75 | |
| GenBank mRNA | AF450480.1 | - | 5 - 6 | 596 | 75 | |
Set Membership: |
|