Assay Details
| Gene Symbol: GSNAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341437.1 | NP_001328932.1 | 5 - 6 | 732 | 82 | |
| NM_118160.5 | NP_567600.1 | 5 - 6 | 626 | 82 | ||
| GenBank mRNA | AF177990.1 | - | 5 - 6 | 490 | 82 | |
| AK317666.1 | - | 5 - 6 | 553 | 82 | ||
| BT024855.1 | - | 5 - 6 | 490 | 82 | ||
More Information
Set Membership: |
|

GSNAP
Genomic Map