Assay Details
| Gene Symbol: GSNAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341437.1 | NP_001328932.1 | 6 - 7 | 816 | 82 | |
| NM_118160.5 | NP_567600.1 | 6 - 7 | 710 | 82 | ||
| GenBank mRNA | AF177990.1 | - | 6 - 7 | 574 | 82 | |
| AK317666.1 | - | 6 - 7 | 637 | 82 | ||
| BT024855.1 | - | 6 - 7 | 574 | 82 | ||
More Information
Set Membership: |
|

GSNAP
Genomic Map