Assay Details
| Gene Symbol: GSNAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341437.1 | NP_001328932.1 | 7 - 8 | 901 | 119 | |
| NM_118160.5 | NP_567600.1 | 7 - 8 | 795 | 119 | ||
| GenBank mRNA | AF177990.1 | - | 7 - 8 | 659 | 119 | |
| AK317666.1 | - | 7 - 8 | 722 | 119 | ||
| BT024855.1 | - | 7 - 8 | 659 | 119 | ||
More Information
Set Membership: |
|

GSNAP
Genomic Map