Assay Details
| Gene Symbol: GHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341467.1 | NP_001320016.1 | 1 - 2 | 1002 | 86 | |
| GenBank mRNA | FJ708750.1 | - | 1 - 2 | 1002 | 86 | |
More Information
Set Membership: |
|

GHR1
Genomic Map