Assay Details
| Gene Symbol: DDB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118228.4 | NP_193842.1 | 11 - 12 | 1263 | 62 | |
| GenBank mRNA | AK229805.1 | - | 11 - 12 | 1078 | 62 | |
More Information
Set Membership: |
|

DDB1B
Genomic Map