Assay Details
| Gene Symbol: DDB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118228.4 | NP_193842.1 | 8 - 9 | 847 | 102 | |
| GenBank mRNA | AK229805.1 | - | 8 - 9 | 662 | 102 | |
More Information
Set Membership: |
|

DDB1B
Genomic Map