Assay Details
| Gene Symbol: emb1417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118238.4 | NP_567622.1 | 4 - 5 | 425 | 136 | |
| GenBank mRNA | AK175548.1 | - | 4 - 5 | 417 | 136 | |
| AK176577.1 | - | 4 - 5 | 403 | 136 | ||
| AK176584.1 | - | 4 - 5 | 417 | 136 | ||
| AX765830.1 | - | 3 - 4 | 337 | 136 | ||
| AY084397.1 | - | 4 - 5 | 420 | 136 | ||
More Information
Set Membership: |
|

emb1417
Genomic Map