Assay Details
| Gene Symbol: KT2/3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341531.1 | NP_001328922.1 | 3 - 4 | 1205 | 87 | |
| NM_118342.3 | NP_567651.1 | 3 - 4 | 1205 | 87 | ||
| GenBank mRNA | AK229735.1 | - | 3 - 4 | 684 | 87 | |
| U40154.1 | - | 3 - 4 | 1204 | 87 | ||
More Information
Set Membership: |
|

KT2/3
Genomic Map