Assay Details
| Gene Symbol: KT2/3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341531.1 | NP_001328922.1 | 5 - 6 | 1589 | 66 | |
| NM_118342.3 | NP_567651.1 | 5 - 6 | 1589 | 66 | ||
| GenBank mRNA | AK229735.1 | - | 5 - 6 | 1068 | 66 | |
| U40154.1 | - | 5 - 6 | 1588 | 66 | ||
More Information
Set Membership: |
|

KT2/3
Genomic Map