Assay Details
| Gene Symbol: KT2/3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118342.3 | NP_567651.1 | 7 - 8 | 2342 | 99 | |
| GenBank mRNA | AK229735.1 | - | 7 - 8 | 1821 | 99 | |
| U40154.1 | - | 7 - 8 | 2341 | 99 | ||
More Information
Set Membership: |
|

KT2/3
Genomic Map