Assay Details
| Gene Symbol: TRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118495.4 | NP_194095.2 | 2 - 3 | 555 | 97 | |
| GenBank mRNA | AJ296155.1 | - | 2 - 3 | 393 | 97 | |
| AK226593.1 | - | 2 - 3 | 423 | 97 | ||
More Information
Set Membership: |
|

TRH1
Genomic Map