Assay Details
| Gene Symbol: TRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118495.4 | NP_194095.2 | 4 - 5 | 854 | 80 | |
| GenBank mRNA | AJ296155.1 | - | 4 - 5 | 692 | 80 | |
| AK226593.1 | - | 4 - 5 | 722 | 80 | ||
More Information
Set Membership: |
|

TRH1
Genomic Map