Assay Details
| Gene Symbol: TRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118495.4 | NP_194095.2 | 5 - 6 | 1117 | 72 | |
| GenBank mRNA | AJ296155.1 | - | 5 - 6 | 955 | 72 | |
| AK226593.1 | - | 5 - 6 | 985 | 72 | ||
More Information
Set Membership: |
|

TRH1
Genomic Map