Assay Details
| Gene Symbol: MGT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124787.1 | NP_001118259.1 | 3 - 4 | 990 | 73 | |
| NM_126412.6 | NP_178460.1 | 2 - 3 | 664 | 73 | ||
| GenBank mRNA | AF499434.1 | - | 1 - 2 | 453 | 73 | |
| AY070380.1 | - | 2 - 3 | 585 | 73 | ||
| AY096567.1 | - | 1 - 2 | 442 | 73 | ||
More Information
Set Membership: |
|

MGT3
Genomic Map