Assay Details
| Gene Symbol: MUR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084163.1 | NP_001077632.1 | 8 - 9 | 1253 | 58 | |
| NM_001332913.1 | NP_001319115.1 | 8 - 9 | 1069 | 58 | ||
| NM_001332914.1 | NP_001321845.1 | 8 - 9 | 1334 | 58 | ||
| NM_001332916.1 | NP_001321844.1 | 7 - 8 | 1125 | 58 | ||
| NM_102799.6 | NP_174350.2 | 8 - 9 | 1288 | 58 | ||
| GenBank mRNA | AF325108.1 | - | 7 - 8 | 865 | 58 | |
| AY064973.1 | - | 8 - 9 | 1201 | 58 | ||
| AY195742.1 | - | 7 - 8 | 903 | 58 | ||
| BT002614.1 | - | 7 - 8 | 865 | 58 | ||
More Information
Set Membership: |
|

MUR4
Genomic Map