Assay Details
| Gene Symbol: RCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160916.1 | NP_001154388.1 | 4 - 5 | 1855 | 80 | |
| NM_001332998.1 | NP_001320331.1 | 4 - 5 | 2362 | 80 | ||
| NM_001332999.1 | NP_001320332.1 | 4 - 5 | 2207 | 80 | ||
| NM_001333000.1 | NP_001320330.1 | 4 - 5 | 2263 | 80 | ||
| NM_102956.3 | NP_564391.1 | 4 - 5 | 1855 | 80 | ||
| NM_179408.3 | NP_849739.1 | 4 - 5 | 2082 | 80 | ||
| GenBank mRNA | AY059796.1 | - | 4 - 5 | 1833 | 80 | |
| AY075633.1 | - | 4 - 5 | 1835 | 80 | ||
| AY142655.1 | - | 2 - 3 | 1367 | 80 | ||
More Information
Set Membership: |
|

RCD1
Genomic Map