Assay Details
| Gene Symbol: RCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332998.1 | NP_001320331.1 | 7 - 8 | 2703 | 61 | |
| NM_001332999.1 | NP_001320332.1 | 7 - 8 | 2548 | 61 | ||
| NM_001333000.1 | NP_001320330.1 | 7 - 8 | 2604 | 61 | ||
| NM_102956.3 | NP_564391.1 | 7 - 8 | 2199 | 61 | ||
| NM_179408.3 | NP_849739.1 | 7 - 8 | 2423 | 61 | ||
| GenBank mRNA | AY059796.1 | - | 7 - 8 | 2174 | 61 | |
| AY075633.1 | - | 7 - 8 | 2179 | 61 | ||
| AY142655.1 | - | 5 - 6 | 1708 | 61 | ||
More Information
Set Membership: |
|

RCD1
Genomic Map