Assay Details
| Gene Symbol: AT4G38480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120009.3 | NP_568042.1 | 1 - 2 | 223 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G38480| Gene Symbol: AT4G38480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120009.3 | NP_568042.1 | 1 - 2 | 223 | 67 | |
Set Membership: |
|