Assay Details
| Gene Symbol: FBA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120057.4 | NP_568049.1 | 1 - 2 | 525 | 72 | |
| NM_202981.1 | NP_974710.1 | 1 - 2 | 257 | 72 | ||
| GenBank mRNA | AK226247.1 | - | 1 - 2 | 258 | 72 | |
| BT015870.1 | - | 1 - 2 | 176 | 72 | ||
More Information
Set Membership: |
|

FBA2
Genomic Map