Assay Details
| Gene Symbol: NST-K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204038.1 | NP_001190967.1 | 3 - 4 | 487 | 84 | |
| NM_120099.4 | NP_568059.1 | 2 - 3 | 474 | 84 | ||
| NM_179196.2 | NP_849527.1 | 2 - 3 | 452 | 84 | ||
| GenBank mRNA | AY091057.1 | - | 2 - 3 | 366 | 84 | |
| AY142535.1 | - | 1 - 2 | 143 | 84 | ||
| KJ667160.1 | - | 1 - 2 | 143 | 84 | ||
| KP872771.1 | - | 1 - 2 | 143 | 84 | ||
More Information
Set Membership: |
|

NST-K1
Genomic Map