Assay Details
| Gene Symbol: NST-K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204038.1 | NP_001190967.1 | 5 - 6 | 872 | 149 | |
| NM_120099.4 | NP_568059.1 | 4 - 5 | 859 | 149 | ||
| NM_179196.2 | NP_849527.1 | 4 - 5 | 837 | 149 | ||
| GenBank mRNA | AY091057.1 | - | 4 - 5 | 751 | 149 | |
| AY142535.1 | - | 3 - 4 | 528 | 149 | ||
| KJ667160.1 | - | 3 - 4 | 528 | 149 | ||
| KP872771.1 | - | 3 - 4 | 528 | 149 | ||
More Information
Set Membership: |
|

NST-K1
Genomic Map