Assay Details
| Gene Symbol: ATHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127118.4 | NP_179159.1 | 1 - 2 | 280 | 64 | |
| GenBank mRNA | AK176140.1 | - | 1 - 2 | 235 | 64 | |
| BT025735.1 | - | 1 - 2 | 204 | 64 | ||
More Information
Set Membership: |
|

ATHM3
Genomic Map