Assay Details
| Gene Symbol: IRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127306.4 | NP_565419.1 | 2 - 3 | 424 | 64 | |
| GenBank mRNA | AB049937.1 | - | 1 - 2 | 156 | 64 | |
| AF308596.1 | - | 2 - 3 | 278 | 64 | ||
More Information
Set Membership: |
|

IRE1A
Genomic Map