Assay Details
| Gene Symbol: AT2G17580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127312.3 | NP_179349.2 | 13 - 14 | 1538 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G17580| Gene Symbol: AT2G17580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127312.3 | NP_179349.2 | 13 - 14 | 1538 | 79 | |
Set Membership: |
|