Assay Details
| Gene Symbol: MCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036289.1 | NP_001031366.1 | 4 - 5 | 963 | 127 | |
| NM_001036290.2 | NP_001031367.1 | 4 - 5 | 1062 | 127 | ||
| NM_001036291.1 | NP_001031368.1 | 4 - 5 | 963 | 127 | ||
| NM_001335556.1 | NP_001324196.1 | 3 - 4 | 990 | 127 | ||
| NM_001335557.1 | NP_001324198.1 | 3 - 4 | 1099 | 127 | ||
| NM_001335558.1 | NP_001324199.1 | 2 - 3 | 1354 | 127 | ||
| NM_001335559.1 | NP_001324200.1 | 3 - 4 | 1277 | 127 | ||
| NM_001335560.1 | NP_001324197.1 | 3 - 4 | 1240 | 127 | ||
| NM_001335561.1 | NP_001324195.1 | 3 - 4 | 1240 | 127 | ||
| NM_127332.3 | NP_179369.2 | 4 - 5 | 1062 | 127 | ||
| GenBank mRNA | AB196961.1 | - | 2 - 3 | 813 | 127 | |
More Information
Set Membership: |
|

MCA2
Genomic Map