Assay Details
| Gene Symbol: TPR15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336923.1 | NP_001324857.1 | 4 - 5 | 2235 | 90 | |
| NM_180020.4 | NP_850351.1 | 4 - 5 | 2240 | 90 | ||
| NM_201930.1 | NP_973659.1 | 4 - 5 | 2222 | 90 | ||
More Information
Set Membership: |
|

TPR15
Genomic Map