Assay Details
| Gene Symbol: EMB2654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336935.1 | NP_001323745.1 | 5 - 6 | 2036 | 69 | |
| NM_180025.2 | NP_850356.1 | 5 - 6 | 2036 | 69 | ||
| NM_180026.2 | NP_850357.2 | 4 - 5 | 1516 | 69 | ||
| GenBank mRNA | AY091135.1 | - | 5 - 6 | 1846 | 69 | |
| AY117242.1 | - | 5 - 6 | 1838 | 69 | ||
More Information
Set Membership: |
|

EMB2654
Genomic Map