Assay Details
| Gene Symbol: KIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125074.2 | NP_001118546.1 | 10 - 11 | 1573 | 69 | |
| NM_110974.5 | NP_566130.1 | 10 - 11 | 1689 | 69 | ||
| NM_180157.1 | NP_850488.1 | 10 - 11 | 1831 | 69 | ||
| GenBank mRNA | AY093170.1 | - | 10 - 11 | 1831 | 69 | |
| BT010386.1 | - | 10 - 11 | 1487 | 69 | ||
| DQ778957.1 | - | 9 - 10 | 1418 | 69 | ||
| X94757.1 | - | 1547 | 69 | |||
More Information
Set Membership: |
|

KIN10
Genomic Map