Assay Details
| Gene Symbol: SGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337686.1 | NP_001326118.1 | 5 - 6 | 1462 | 80 | |
| NM_111582.3 | NP_566297.2 | 6 - 7 | 1363 | 80 | ||
| NM_180198.3 | NP_850529.1 | 6 - 7 | 1274 | 80 | ||
| GenBank mRNA | AK316817.1 | - | 6 - 7 | 1079 | 80 | |
| AY057496.1 | - | 6 - 7 | 920 | 80 | ||
| AY079032.1 | - | 5 - 6 | 751 | 80 | ||
| KJ138837.1 | - | 6 - 7 | 997 | 80 | ||
More Information
Set Membership: |
|

SGT
Genomic Map