Assay Details
| Gene Symbol: MYB4R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202993.1 | NP_001189922.1 | 4 - 5 | 960 | 69 | |
| NM_001338329.1 | NP_001327710.1 | 3 - 4 | 1097 | 69 | ||
| NM_112692.3 | NP_566597.1 | 2 - 3 | 1065 | 69 | ||
| NM_180276.2 | NP_850607.1 | 3 - 4 | 995 | 69 | ||
| GenBank mRNA | AY033827.1 | - | 2 - 3 | 939 | 69 | |
More Information
Set Membership: |
|

MYB4R1
Genomic Map