Assay Details
| Gene Symbol: MYB4R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202993.1 | NP_001189922.1 | 8 - 9 | 2130 | 65 | |
| NM_001338329.1 | NP_001327710.1 | 7 - 8 | 2267 | 65 | ||
| NM_112692.3 | NP_566597.1 | 6 - 7 | 2235 | 65 | ||
| NM_180276.2 | NP_850607.1 | 7 - 8 | 2165 | 65 | ||
| GenBank mRNA | AY033827.1 | - | 6 - 7 | 2109 | 65 | |
More Information
Set Membership: |
|

MYB4R1
Genomic Map