Assay Details
| Gene Symbol: ARC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112862.4 | NP_188606.2 | 8 - 9 | 1421 | 60 | |
| NM_180284.3 | NP_850615.2 | 8 - 9 | 1421 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARC5| Gene Symbol: ARC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112862.4 | NP_188606.2 | 8 - 9 | 1421 | 60 | |
| NM_180284.3 | NP_850615.2 | 8 - 9 | 1421 | 60 | ||
Set Membership: |
|