Assay Details
| Gene Symbol: GIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160826.1 | NP_001154298.1 | 4 - 5 | 606 | 69 | |
| NM_099998.4 | NP_563619.1 | 3 - 4 | 522 | 69 | ||
| GenBank mRNA | AF410307.1 | - | 3 - 4 | 442 | 69 | |
| AY102133.1 | - | 3 - 4 | 278 | 69 | ||
| AY102640.1 | - | 3 - 4 | 482 | 69 | ||
More Information
Set Membership: |
|

GIF2
Genomic Map