Assay Details
| Gene Symbol: AGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120126.5 | NP_568064.1 | 1 - 2 | 400 | 69 | |
| GenBank mRNA | AK227118.1 | - | 1 - 2 | 262 | 69 | |
| AY054264.1 | - | 1 - 2 | 254 | 69 | ||
| BT002306.1 | - | 1 - 2 | 230 | 69 | ||
More Information
Set Membership: |
|

AGT2
Genomic Map