Assay Details
| Gene Symbol: Fes1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036745.2 | NP_001031822.1 | 2 - 3 | 222 | 65 | |
| NM_001342648.1 | NP_001332564.1 | 3 - 4 | 421 | 65 | ||
| NM_120293.3 | NP_195835.1 | 2 - 3 | 277 | 65 | ||
| GenBank mRNA | AK118831.1 | - | 2 - 3 | 240 | 65 | |
| BT005554.1 | - | 2 - 3 | 229 | 65 | ||
More Information
Set Membership: |
|

Fes1C
Genomic Map