Assay Details
| Gene Symbol: DPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120325.4 | NP_195867.2 | 3 - 4 | 520 | 122 | |
| NM_180696.2 | NP_851027.1 | 4 - 5 | 329 | 122 | ||
| NM_180697.3 | NP_851028.1 | 4 - 5 | 475 | 122 | ||
| GenBank mRNA | AJ294531.1 | - | 4 - 5 | 401 | 122 | |
| AJ319027.1 | - | 3 - 4 | 306 | 122 | ||
| AK117135.1 | - | 4 - 5 | 291 | 122 | ||
| BT005286.1 | - | 3 - 4 | 228 | 122 | ||
More Information
Set Membership: |
|

DPA
Genomic Map