Assay Details
| Gene Symbol: DPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120325.4 | NP_195867.2 | 6 - 7 | 801 | 103 | |
| NM_180696.2 | NP_851027.1 | 7 - 8 | 610 | 103 | ||
| NM_180697.3 | NP_851028.1 | 7 - 8 | 756 | 103 | ||
| GenBank mRNA | AJ294531.1 | - | 7 - 8 | 682 | 103 | |
| AJ319027.1 | - | 6 - 7 | 587 | 103 | ||
| AK117135.1 | - | 7 - 8 | 572 | 103 | ||
| BT005286.1 | - | 6 - 7 | 509 | 103 | ||
More Information
Set Membership: |
|

DPA
Genomic Map