Assay Details
| Gene Symbol: HD2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120455.4 | NP_195994.3 | 2 - 3 | 214 | 97 | |
| GenBank mRNA | AF255712.1 | - | 1 - 2 | 79 | 97 | |
| AF372889.1 | - | 2 - 3 | 191 | 97 | ||
| AF510168.1 | - | 1 - 2 | 79 | 97 | ||
| AY142039.1 | - | 1 - 2 | 79 | 97 | ||
More Information
Set Membership: |
|

HD2C
Genomic Map